KEY497445053 Key Organics / BIONET

ethyl (2Z)-3-amino-4,4,4-trifluorobut-2-enoate

  • MDL number MFCD00041541
  • Cas Number : 372-29-2
  • Molecular Weight : 183.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/654947

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9F-023_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart