KEY497445055 Key Organics / BIONET

2-hydroxy-9-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD00749757
  • Cas Number : 17326-09-9
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/654955

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3233_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart