KEY508730417 Key Organics / BIONET

10,20-dimethoxyhexacyclo[17.8.0.0^{2,11}.0^{3,8}.0^{13,17}.0^{22,27}]heptacosa-1(19),2,4,6,8,10,13(17),15,20,22,24,26-dodecaene

  • MDL number MFCD29918522
  • Cas Number : 1417714-08-9
  • Molecular Weight : 404.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/655871

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-00052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart