KEY508730439 Key Organics / BIONET

methyl 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD17011994
  • Cas Number : 1321613-00-6
  • Molecular Weight : 298.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Solid
Iupac Name methyl 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Inchi Code 1S/C14H17BF2O4/c1-13(2)14(3,4)21-15(20-13)8-6-9(16)11(10(17)7-8)12(18)19-5/h6-7H,1-5H3
Inchi Key DLAZQBFMFDOBGT-UHFFFAOYSA-N

Description

Melting Point Solid
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/655209

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3251_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart