KEY508730475 Key Organics / BIONET

1,4-dimethyl (2E)-2,3-bis(acetyloxy)but-2-enedioate

  • MDL number MFCD01726213
  • Cas Number : 130-84-7
  • Molecular Weight : 260.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/655211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3253_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart