KEY514452708 Key Organics / BIONET

6-bromo-5-methylpyridine-3-carbonitrile

  • MDL number MFCD03095063
  • Cas Number : 374633-37-1
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-5-methylnicotinonitrile
Inchi Code 1S/C7H5BrN2/c1-5-2-6(3-9)4-10-7(5)8/h2,4H,1H3
Inchi Key JECOSEMUVCECAL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/691449

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-4094_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart