KEY514452818 Key Organics / BIONET

(1H-pyrazol-1-yl)methanol

  • MDL number MFCD00955584
  • Cas Number : 1120-82-7
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-pyrazol-1-ylmethanol
Inchi Code 1S/C4H6N2O/c7-4-6-3-1-2-5-6/h1-3,7H,4H2
Inchi Key BYUMAPPWWKNLNX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/691559

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-4206_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart