KEY514453136 Key Organics / BIONET

2-(2,4-dichlorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD27935406
  • Cas Number : 90389-07-4
  • Molecular Weight : 226.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/691876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-4525_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart