KEY514453562 Key Organics / BIONET

tert-butyl N-[2-(2-aminoethoxy)ethyl]carbamate

  • MDL number MFCD12031501
  • Cas Number : 127828-22-2
  • Molecular Weight : 7/22/1900 6:28:48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/692303

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-5737_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart