KEY514454011 Key Organics / BIONET

tert-butyl 3-[2-(2-aminoethoxy)ethoxy]propanoate

  • MDL number MFCD11041135
  • Cas Number : 756525-95-8
  • Molecular Weight : 233.308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Store at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/692716

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-6156_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart