KEY514454224 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoate

  • MDL number MFCD09952636
  • Cas Number : 120550-35-8
  • Molecular Weight : 410.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/692930

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-6375_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart