KEY514454331 Key Organics / BIONET

4-(dimethoxymethyl)pyrimidin-2-amine

  • MDL number MFCD05864792
  • Cas Number : 165807-05-6
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(dimethoxymethyl)-2-pyrimidinamine
Inchi Code 1S/C7H11N3O2/c1-11-6(12-2)5-3-4-9-7(8)10-5/h3-4,6H,1-2H3,(H2,8,9,10)
Inchi Key OQINWXZQCAIVNK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/693037

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-6483_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart