KEY514454778 Key Organics / BIONET

4-(2-chloro-6-phenylpyrimidin-4-yl)morpholine

  • MDL number MFCD30183265
  • Cas Number : 54994-38-6
  • Molecular Weight : 275.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1041524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4959_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart