KEY514454779 Key Organics / BIONET

(1R,3S)-3-aminocyclopentan-1-ol

  • MDL number MFCD11052498
  • Cas Number : 1110772-05-8
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at -18
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/693477

Safety Information

signalword Warning
pcodes P232,P260,P270,P280,P402 + P404,P411,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4960_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart