KEY771305504 Key Organics / BIONET

(3S)-3-amino-3-(4-aminophenyl)propan-1-ol hydrochloride

  • MDL number MFCD30717717
  • Cas Number : 1374669-67-6
  • Molecular Weight : 202.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665443

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart