KEY771305511 Key Organics / BIONET

ethyl (2S,3R)-3-hydroxy-1-(4-methoxyphenyl)-3-methyl-2-phenylpyrrolidine-2-carboxylate

  • MDL number MFCD30717727
  • Cas Number : 2098497-05-1
  • Molecular Weight : 355.434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Shipping Temperature Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665621

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00065_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart