KEY771305512 Key Organics / BIONET

{imidazo[1,2-b]pyridazin-2-yl}methanol

  • MDL number MFCD30717728
  • Cas Number : 1487220-02-9
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4940_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart