KEY771305516 Key Organics / BIONET

2-amino-2-(2,4-dichlorophenyl)ethan-1-ol

  • MDL number MFCD12782009
  • Cas Number : 1184839-78-8
  • Molecular Weight : 206.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-2-(2,4-dichlorophenyl)ethanol
Inchi Code 1S/C8H9Cl2NO/c9-5-1-2-6(7(10)3-5)8(11)4-12/h1-3,8,12H,4,11H2
Inchi Key VWMOCFARWTYBNT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665320

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9727_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart