KEY771305521 Key Organics / BIONET

2-amino-2-(3-chlorophenyl)ethan-1-ol

  • MDL number MFCD12827190
  • Cas Number : 179811-63-3
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-2-(3-chlorophenyl)ethanol
Inchi Code 1S/C8H10ClNO/c9-7-3-1-2-6(4-7)8(10)5-11/h1-4,8,11H,5,10H2
Inchi Key HLXHAXFWWGYXQW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665325

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9732_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart