KEY771305526 Key Organics / BIONET

tert-butyl N-[2-amino-1-(2-ethylphenyl)ethyl]carbamate

  • MDL number MFCD17337968
  • Cas Number : 1270803-91-2
  • Molecular Weight : 264.369

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665330

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9737_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart