KEY771305528 Key Organics / BIONET

8-methoxy-2,3,4,5-tetrahydro-1,4-benzoxazepine

  • MDL number MFCD14582632
  • Cas Number : 402933-49-7
  • Molecular Weight : 179.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665332

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9739_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart