KEY771305545 Key Organics / BIONET

2-methyl-1,3-diazaspiro[4.4]non-1-en-4-one

  • MDL number MFCD04037196
  • Cas Number : 759442-73-4
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665349

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9756_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart