KEY771305546 Key Organics / BIONET

2-amino-4-(morpholin-4-yl)butanoic acid

  • MDL number MFCD07786927
  • Cas Number : 770684-35-0
  • Molecular Weight : 188.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665350

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9757_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart