KEY771305547 Key Organics / BIONET

3-amino-1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid

  • MDL number MFCD03790945
  • Cas Number : 368866-17-5
  • Molecular Weight : 244.291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665351

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-9758_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart