KEY771305554 Key Organics / BIONET

2-(4-bromothiophen-2-yl)-2-(pyrrolidin-1-yl)acetic acid

  • MDL number MFCD14632477
  • Cas Number : 1249997-23-6
  • Molecular Weight : 290.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9765_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart