KEY771305555 Key Organics / BIONET

3-methyl-2-[(2,2,2-trifluoroethyl)amino]butanamide

  • MDL number MFCD14619024
  • Cas Number : 1247717-47-0
  • Molecular Weight : 198.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665358

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9766_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart