KEY771305572 Key Organics / BIONET

2-amino-2-ethyl-3-methylbutanamide

  • MDL number MFCD12817049
  • Cas Number : 1179897-71-2
  • Molecular Weight : 144.218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Colourless to Off-white Low Melting Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665375

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-9784_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart