KEY771305574 Key Organics / BIONET

8-bromo-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD09261262
  • Cas Number : 688363-48-6
  • Molecular Weight : 228.045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart