KEY771305803 Key Organics / BIONET

(1S)-1-(5-bromopyridin-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD18711631
  • Cas Number : 1391450-63-7
  • Molecular Weight : 273.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-1-(5-bromo-2-pyridinyl)ethanamine dihydrochloride
Inchi Code 1S/C7H9BrN2.2ClH/c1-5(9)7-3-2-6(8)4-10-7;;/h2-5H,9H2,1H3;2*1H/t5-;;/m0../s1
Inchi Key SOOAYJPBXRUXBZ-XRIGFGBMSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665387

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7110_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart