KEY771305805 Key Organics / BIONET

(1R)-1-(2-methylphenyl)propan-1-amine hydrochloride

  • MDL number MFCD11101147
  • Cas Number : 856562-93-1
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(2-methylphenyl)-1-propanamine hydrochloride
Inchi Code 1S/C10H15N.ClH/c1-3-10(11)9-7-5-4-6-8(9)2;/h4-7,10H,3,11H2,1-2H3;1H/t10-;/m1./s1
Inchi Key ZRFSFSDKCOAXNJ-HNCPQSOCSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665389

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7113_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart