KEY771305806 Key Organics / BIONET

(1S)-5-bromo-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD26585976
  • Cas Number : 1810074-82-8
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-5-bromo-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
Inchi Code 1S/C10H12BrN.ClH/c11-9-5-1-4-8-7(9)3-2-6-10(8)12;/h1,4-5,10H,2-3,6,12H2;1H/t10-;/m0./s1
Inchi Key OGJCTIOLCPDGAX-PPHPATTJSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665390

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7115_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart