KEY771305808 Key Organics / BIONET

(1R)-1-(5-fluoropyridin-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD29060310
  • Cas Number : 1909287-34-8
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665392

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7119_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart