KEY771305810 Key Organics / BIONET

(2R)-2-amino-2-(naphthalen-2-yl)ethan-1-ol hydrochloride

  • MDL number MFCD12758444
  • Cas Number : 204851-80-9
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7121_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart