KEY771305813 Key Organics / BIONET

(2R)-2-amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid hydrochloride

  • MDL number MFCD28986153
  • Cas Number : 1810074-80-6
  • Molecular Weight : 285.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-2-amino-3-(4-(trifluoromethoxy)phenyl)propanoic acid hydrochloride
Inchi Code 1S/C10H10F3NO3.ClH/c11-10(12,13)17-7-3-1-6(2-4-7)5-8(14)9(15)16;/h1-4,8H,5,14H2,(H,15,16);1H/t8-;/m1./s1
Inchi Key UAKYDNOGIWVOQQ-DDWIOCJRSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665397

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7127_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart