KEY771305832 Key Organics / BIONET

(4S)-6-fluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD15474861
  • Cas Number : 1260609-97-9
  • Molecular Weight : 203.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4S)-6-fluoro-3,4-dihydro-2H-chromen-4-amine hydrochloride
Inchi Code 1S/C9H10FNO.ClH/c10-6-1-2-9-7(5-6)8(11)3-4-12-9;/h1-2,5,8H,3-4,11H2;1H/t8-;/m0./s1
Inchi Key VHMBQKMHABXVJN-QRPNPIFTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665416

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7166_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart