KEY771305834 Key Organics / BIONET

(1S)-1-(5-methylfuran-2-yl)propan-1-amine hydrochloride

  • MDL number MFCD24423898
  • Cas Number : 1810074-74-8
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7170_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart