KEY771305835 Key Organics / BIONET

(2R)-2-[4-(trifluoromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD08751409
  • Cas Number : 1391407-62-7
  • Molecular Weight : 251.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-2-[4-(trifluoromethyl)phenyl]pyrrolidine hydrochloride
Inchi Code 1S/C11H12F3N.ClH/c12-11(13,14)9-5-3-8(4-6-9)10-2-1-7-15-10;/h3-6,10,15H,1-2,7H2;1H/t10-;/m1./s1
Inchi Key XHVVDNXOJJAMDE-HNCPQSOCSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665419

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7172_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart