KEY771305837 Key Organics / BIONET

(2S)-2-(2-methoxyphenyl)piperidine hydrochloride

  • MDL number MFCD12910640
  • Cas Number : 1391417-03-0
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-(2-methoxyphenyl)piperidine hydrochloride
Inchi Code 1S/C12H17NO.ClH/c1-14-12-8-3-2-6-10(12)11-7-4-5-9-13-11;/h2-3,6,8,11,13H,4-5,7,9H2,1H3;1H/t11-;/m0./s1
Inchi Key PUSGLZAYZOPJCB-MERQFXBCSA-N

Description

Purity 95%
Country of Origin GB
Availability details for
Quantity
  • Availability
  • Add to Cart