KEY771305839 Key Organics / BIONET

(4S)-4-amino-4-phenylbutan-1-ol hydrochloride

  • MDL number MFCD24431191
  • Cas Number : 1810074-83-9
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665423

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7177_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart