KEY771305844 Key Organics / BIONET

(1R)-1-{4-[(trifluoromethyl)sulfanyl]phenyl}ethan-1-amine hydrochloride

  • MDL number MFCD24418250
  • Cas Number : 1800240-39-4
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(4-((trifluoromethyl)thio)phenyl)ethan-1-amine hydrochloride
Inchi Code 1S/C9H10F3NS.ClH/c1-6(13)7-2-4-8(5-3-7)14-9(10,11)12;/h2-6H,13H2,1H3;1H/t6-;/m1./s1
Inchi Key FATBLICFSRIJRR-FYZOBXCZSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665428

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7182_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart