KEY771305846 Key Organics / BIONET

(1R)-1-(4-fluorophenyl)but-3-en-1-amine hydrochloride

  • MDL number MFCD12068566
  • Cas Number : 1391361-52-6
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(4-fluorophenyl)-3-buten-1-amine hydrochloride
Inchi Code 1S/C10H12FN.ClH/c1-2-3-10(12)8-4-6-9(11)7-5-8;/h2,4-7,10H,1,3,12H2;1H/t10-;/m1./s1
Inchi Key TXYCZYDHHPXNIK-HNCPQSOCSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665430

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7185_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart