KEY771305853 Key Organics / BIONET

(1R)-1-[3-chloro-4-(trifluoromethyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD24419924
  • Cas Number : 1810074-87-3
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665437

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7198_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart