KEY771305857 Key Organics / BIONET

3,4-diethyl (3S,4S)-1-benzylpyrrolidine-3,4-dicarboxylate

  • MDL number MFCD23136921
  • Cas Number : 112794-79-3
  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/664885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0226_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart