KEY771305861 Key Organics / BIONET

methyl 3-[(1R)-1-amino-2-hydroxyethyl]benzoate hydrochloride

  • MDL number MFCD12910850
  • Cas Number : 1391515-70-0
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-[(1R)-1-amino-2-hydroxyethyl]benzoate hydrochloride
Inchi Code 1S/C10H13NO3.ClH/c1-14-10(13)8-4-2-3-7(5-8)9(11)6-12;/h2-5,9,12H,6,11H2,1H3;1H/t9-;/m0./s1
Inchi Key SWIJKZSHJUALNL-FVGYRXGTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665445

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7207_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart