KEY771305863 Key Organics / BIONET

(1R)-1-(6-fluoropyridin-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD30717718
  • Cas Number : 2061996-65-2
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(6-fluoropyridin-2-yl)ethan-1-amine dihydrochloride
Inchi Code 1S/C7H9FN2.2ClH/c1-5(9)6-3-2-4-7(8)10-6;;/h2-5H,9H2,1H3;2*1H/t5-;;/m1../s1
Inchi Key LAPSSXQERAHMRK-ZJIMSODOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665447

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7209_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart