KEY771305865 Key Organics / BIONET

8-[(1S,2S,6R,8S)-6,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0^{2,6}]decan-4-yl]quinoline

  • MDL number MFCD30717695
  • Cas Number : 1234575-68-8
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/664887

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3267_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart