KEY771305867 Key Organics / BIONET

(1S)-1-(4-chloro-2-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD16294095
  • Cas Number : 1213908-11-2
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-1-(4-chloro-2-methylphenyl)ethanamine hydrochloride
Inchi Code 1S/C9H12ClN.ClH/c1-6-5-8(10)3-4-9(6)7(2)11;/h3-5,7H,11H2,1-2H3;1H/t7-;/m0./s1
Inchi Key FAAKQSKVORTCAV-FJXQXJEOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665451

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7213_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart