KEY771305943 Key Organics / BIONET

(2S)-2-amino-2-(3,5-difluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD24414217
  • Cas Number : 1212932-15-4
  • Molecular Weight : 209.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665455

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart