KEY771305947 Key Organics / BIONET

(1S)-1-(2-chlorophenyl)propan-1-amine hydrochloride

  • MDL number MFCD12756981
  • Cas Number : 873893-94-8
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7223_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart