KEY771305952 Key Organics / BIONET

(R)-cyclopropyl(3-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD19705372
  • Cas Number : 1212958-89-8
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-cyclopropyl(3-methoxyphenyl)methanamine hydrochloride
Inchi Code 1S/C11H15NO.ClH/c1-13-10-4-2-3-9(7-10)11(12)8-5-6-8;/h2-4,7-8,11H,5-6,12H2,1H3;1H/t11-;/m1./s1
Inchi Key YZEPXOHQBWEEEY-RFVHGSKJSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665464

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7229_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart